Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt
Shop All Red Kap Industries Products
Click to view available options
Size:
2X-Large (Long)
Description
- Lightweight fabric flexes for added comfort and range of movement
- Contrasting pattern on sleeve for an athletic look
- Curved flatlock seams for optimal fit
- Unique front pocket design with pencil stall
- Ribbed collar
- CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Size | 2X-Large (Long) |
| Gender | Men's |
| Material | Cotton, Spandex |
| Color | Navy |
| Neck Style | Hemmed Crew |
| Sleeve Style | Long Sleeve |
| Construction | Knit |
| Closure Type | Button |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction