Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mena Antibody (ENAH/1988), Biotin, Novus Biologicals™
SDP

Catalog No. NB011857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011857 Supplier Novus Biologicals Supplier No. NBP279887B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Mena
Applications Western Blot, ELISA
Classification Monoclonal
Clone ENAH/1988
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog
Gene Symbols ENAH
Host Species Mouse
Immunogen Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) (Uniprot: Q8N8S7)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.