Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menin Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292073 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292073 Supplier Novus Biologicals Supplier No. NB100399APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Menin Polyclonal antibody specifically detects Menin in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Menin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias menin, multiple, multiple endocrine neoplasia I
Host Species Rabbit
Immunogen A synthetic peptide, which represented a portion of human Multiple Endocrine Neoplasia 1 encoded within exon 11 (MEN1 LocusLink ID 4221).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4221
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.