Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menin Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP171794 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171794 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171794 Supplier Novus Biologicals Supplier No. NBP171794

Goat Polyclonal Antibody

Menin Polyclonal specifically detects Menin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Menin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1:1000-1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation PBS, 0.2% BSA, and 20% Trehalose with 0.09% Sodium Azide
Gene Accession No. NP_000235.2
Gene Alias menin, multiple, multiple endocrine neoplasia I
Gene Symbols MEN1
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221).
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4221
Reconstitution Reconstitute with 0.1 ml deionized water.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.