Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menin Antibody, Novus Biologicals™
SDP

Catalog No. NBP217277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217277 Supplier Novus Biologicals Supplier No. NBP217277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Menin Polyclonal specifically detects Menin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Menin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.03mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O00255
Gene Alias menin, multiple, multiple endocrine neoplasia I
Gene Symbols MEN1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Menin. The exact sequence is proprietary.
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4221
Test Specificity Human. Species reactivity based on sequence identity: Canine (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.