Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt
Shop All Red Kap Industries Products
Click to view available options
Size:
X-Large
3X-Large
Description
- Extra comfortable fabric
- Underarm gusset for improved breathability
- Rib-knit crewneck collar
- Single spade chest pocket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Size | 3X-Large |
| Gender | Men's |
| Material | FR Cotton |
| Color | Navy |
| Neck Style | Hemmed Crew |
| Sleeve Style | Long Sleeve |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Hazard Risk Category | Category 2 |
| Arc Flash Rating | 8.9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction