Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menthofuran, 95%

Catalog No. p-3778937
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 494-90-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494-90-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00041851
InChI Key YGWKXXYGDYYFJU-UHFFFAOYSA-N
Synonym menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
PubChem CID 329983
ChEBI CHEBI:50542
IUPAC Name 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
SMILES CC1CCC2=C(C1)OC=C2C

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 87.0°C (15.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4832 to 1.4852
Quantity 10 g
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble
Formula Weight 150.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Menthofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-795-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.