Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEOX 2 Antibody (OTI4F6), PE, Novus Biologicals™
SDP

Catalog No. NB287915 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287915 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287915 Supplier Novus Biologicals Supplier No. NBP272650PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEOX 2 Monoclonal antibody specifically detects MEOX 2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MEOX 2
Applications Western Blot
Classification Monoclonal
Clone OTI4F6
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias growth arrest-specific homeo box, Growth arrest-specific homeobox, homeobox protein MOX-2, mesenchyme homeo box 2 (growth arrest-specific homeo box), mesenchyme homeobox 2GAX, MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box)
Host Species Mouse
Immunogen Full length human recombinant protein of human MEOX2 (NP_005915) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4223
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.