Learn More
MEP50 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | MEP50 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Gene | WDR77 |
| Gene Accession No. | Q9BQA1 |
| Gene Alias | 2610003I18Rik; 2610312E17Rik; Ac2-269; Androgen receptor cofactor p44; C79984; HKMT1069; Mep50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77 |
| Gene Symbols | WDR77 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.