Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEPCE Antibody, Novus Biologicals™
SDP

Catalog No. NB10093420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093420 Supplier Novus Biologicals Supplier No. NB10093420
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MEPCE Polyclonal specifically detects MEPCE in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen MEPCE
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_062552.2
Gene Alias BCDIN37SK snRNA methylphosphate capping enzyme, bicoid-interacting 3, homolog, Bin3 homolog, bin3, bicoid-interacting 3, homolog (Drosophila), EC 2.1.1.-, FLJ20257, methylphosphate capping enzyme
Gene Symbols MEPCE
Host Species Goat
Immunogen Peptide with sequence KQQRKFQYGNYCK corresponding to internal region according to NP_062552.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56257
Test Specificity Western blot: Preliminary testing was unsuccessful on Human for this particular batch.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.