Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meprin alpha Subunit/MEP1A Antibody (017), PE, Novus Biologicals™
SDP

Catalog No. NB308106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308106 Supplier Novus Biologicals Supplier No. NBP289288PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Meprin alpha Subunit/MEP1A Monoclonal antibody specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for ELISA

Specifications

Antigen Meprin alpha Subunit/MEP1A
Applications ELISA
Classification Monoclonal
Clone 017
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Meprin alpha Subunit/MEP1A (Accession#: NP_005579.2; Met1-Gln601).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4224
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.