Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meprin alpha Subunit/MEP1A Antibody (364312) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB372797 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372797 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372797 Supplier Novus Biologicals Supplier No. FAB3220M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Meprin alpha Subunit/MEP1A Monoclonal specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunoprecipitation, Western Blot.

Specifications

Antigen Meprin alpha Subunit/MEP1A
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 364312
Conjugate DyLight 594
Dilution Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A, Val22-Gln601, Accession # AAA21338
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4224
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.