Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meprin beta Subunit/MEP1B Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB083564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB083564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB083564 Supplier Novus Biologicals Supplier No. NBP299477H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Meprin beta Subunit/MEP1B Polyclonal specifically detects Meprin beta Subunit/MEP1B in Human samples. It is validated for ELISA.

Specifications

Antigen Meprin beta Subunit/MEP1B
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias EC 3.4.24.63, endopeptidase-2, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Meprin beta Subunit/MEP1B (NP_005916.2; Met1-Thr652).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4225
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.