Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer Antibody (3D07) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018056 0.025 mg
NB11094148 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018056 Supplier Novus Biologicals Supplier No. NB110941480.025MG

Rat Monoclonal Antibody

Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Mer
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3D07
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1000, Immunohistochemistry-Paraffin 1:50-1:200
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q60805
Gene Alias c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Gene Symbols MERTK
Host Species Rat
Immunogen Recombinant Mouse Mer extracellular domain
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10461
Test Specificity Mouse Mer. It has no crossreactivity with rmAxl or rmDtk.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.