Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer Rabbit anti-Human, Clone: 4I1P7, Novus Biologicals™
SDP

Catalog No. NB165976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165976 100 μg
NB165977 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165976 Supplier Novus Biologicals Supplier No. NBP315440100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Western Blot

Specifications

Antigen Mer
Applications Western Blot
Classification Monoclonal
Clone 4I1P7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Mer (Q12866).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10461
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.