Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog No. p-7050297
Click to view available options
Quantity:
100 g

Catalog No. AA3641922

Supplier: Thermo Scientific Chemicals 03641922

Only null left
Add to Cart

Mercury(I) chloride is used in medicine as a diuretic and purgative, uses in cosmetics as soaps and skin lightening creams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10112-91-1
Molecular Formula Cl2Hg2
Molecular Weight (g/mol) 472.084
MDL Number MFCD00011043
InChI Key ZOMNIUBKTOKEHS-UHFFFAOYSA-L
Synonym mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
PubChem CID 24956
ChEBI CHEBI:33050
IUPAC Name chloromercury
SMILES Cl[Hg].Cl[Hg]

Specifications

Melting Point 400°C (sublimation)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Hg2Cl2
UN Number UN2025
Merck Index 14,5894
Solubility Information Soluble in (0.002g/L )water.
Formula Weight 472.09
Percent Purity ≤99.5%
Grade ACS
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.15 g/mL
Chemical Name or Material Mercury(I) chloride
Show More Show Less
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
EINECSNumber 233-307-5
RTECSNumber OV8740000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.