Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS-CoV/NCov Spike Protein Rabbit anti-MERS-CoV, Alexa Fluor 750, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB142804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB142804 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB142804 Supplier Novus Biologicals Supplier No. NBP305835AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

MERS-CoV/NCov Spike Protein Polyclonal specifically detects MERS-CoV/NCov Spike Protein in MERS-CoV samples. It is validated for Western Blot.

Specifications

Antigen MERS-CoV/NCov Spike Protein
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coronavirus s1, coronavirus s2, coronavirus spike, cov spike, E2, Human coronavirus spike glycoprotein, MERS, MERS Peplomer protein, MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV E2, MERS-CoV Peplomer protein, MERSCoV S glycoprotein, MERS-CoV S glycoprotein, MERSCoV Spike glycoprotein, MERS-CoV Spike glycoprotein, MERSCOV Spike Protein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, ncov RBD, ncov s1, ncov s2, ncov spike, Peplomer proteinMERS-CoV Spike glycoprotein, RBD, S, S1, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, a synthetic peptide corresponding to the S2 subunit of MERS-CoV/NCov Spike Protein (S protein).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14254594
Target Species MERS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.