Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS-CoV/NCov Spike Protein Rabbit anti-MERS-CoV, Clone: 723, Novus Biologicals™
SDP

Catalog No. NB022133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022133 200 μg
NB022134 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022133 Supplier Novus Biologicals Supplier No. NBP290336200UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MERS-CoV/NCov Spike Protein Monoclonal specifically detects MERS-CoV/NCov Spike Protein in MERS-CoV samples. It is validated for Block/Neutralize.

Specifications

Antigen MERS-CoV/NCov Spike Protein
Applications Neutralization
Classification Monoclonal
Clone 723
Conjugate Unconjugated
Formulation 0.2 um filtered solution in PBS
Gene Accession No. AFS88936.1, AFS88936.1
Gene Alias coronavirus s1, coronavirus s2, coronavirus spike, cov spike, ncov RBD, ncov s1, ncov s2, ncov spike, RBD, S, S1, Spike RBD
Host Species Rabbit
Immunogen MERS-CoV (NCoV/ Novel coronavirus) Spike Protein (ECD, aa 1-1297)
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species MERS-CoV
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.