Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS-CoV Nucleoprotein Rabbit anti-MERS-CoV, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167209 100 μg
NB167210 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167209 Supplier Novus Biologicals Supplier No. NBP316057100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MERS-CoV Nucleoprotein Polyclonal antibody specifically detects MERS-CoV Nucleoprotein in MERS-CoV samples. It is validated for Western Blot

Specifications

Antigen MERS-CoV Nucleoprotein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias MERS, MERS NC, MERS Nucleocapsid protein, MERS Nucleoprotein, MERSCoV NC, MERS-CoV NC, MERSCoV Nucleocapsid protein, MERS-CoV Nucleocapsid protein, MERSCOV Nucleoprotein, MERS-CoV Protein N, Middle East respiratory syndrome-related coronavirus, Nucleocapsid protein, Protein N
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-413 of coronavirus Nucleoprotein (YP_009047211.1). MASPAAPRAVSFADNNDITNTNLSRGRGRNPKPRAAPNNTVSWYTGLTQHGKVPLTFPPGQGVPLNANSTPAQNAGYWRRQDRKINTGNGIKQLAPRWYFYYTGTGPEAALPFRAVKDGIVWVHEDGATDAPSTFGTRNPNNDSAIVTQFAPGTKLPKNFHIEGTGGNSQSSSRASSLSRNSSRSSSQGSRSGNSTRGTSPGPSGIGAVGGDLLYLDLLNRLQALESGKVKQSQPKVITKKDAAAAKNKMRHKRTSTKSFNMVQAFGLRGPGDLQGNFGDLQLNKLGTEDPRWPQIAELAPTASAFMGMSQFKLTHQNNDDHGNPVYFLRYSGAIKLDPKNPNYNKWLELLEQNIDAYKTFPKKEKKQKAPKEESTDQMSEPPKEQRVQGSITQRTRTRPSVQPGPMIDVNTD
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 37616439
Target Species MERS-CoV
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.