Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES Monohydrate, ≥98.5% (Dry Basis), Ultrapure

Catalog No. p-7082848
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Melting Point ∼308°C (decomposition)
Packaging Plastic bottle
Linear Formula HO3S(CH2)2(C4H8NO)·H2O
Quantity 100 g
Beilstein 141319
Formula Weight 213.25
Percent Purity ≥98.5%
Grade Ultrapure
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material MES Monohydrate
Show More Show Less
EINECSNumber 224-632-3
RTECSNumber QE3479500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.