Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
200 g
500 g
Packaging:
Poly Bottle
Description
- For Liquid Chromatography and Molecular Biology Applications
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 200 g |
Formula Weight | 213.25 |
Grade | Ultrapure Bioreagent |
Physical Form | Crystals |
Chemical Name or Material | MES, Monohydrate, Free Acidline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction