Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Erythritol, 99%

Catalog No. aaa1581314
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol

meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

Solubility
Soluble in water (0.1 g/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-32-6
Molecular Formula C4H10O4
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004710
InChI Key UNXHWFMMPAWVPI-UHFFFAOYNA-N
Synonym erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
PubChem CID 222285
ChEBI CHEBI:17113
IUPAC Name butane-1,2,3,4-tetrol
SMILES OCC(O)C(O)CO

Specifications

Melting Point 119°C to 124°C
Boiling Point 329°C to 331°C
Assay Percent Range 99%
Quantity 25 g
Beilstein 1719753
Merck Index 14,3675
Solubility Information Soluble in water (0.1g/ml).
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material meso-Erythritol
EINECSNumber 205-737-3
RTECSNumber KF2000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.