Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Erythritol 99.0+%, TCI America™
SDP

Catalog No. E0021100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0021100G 100 g
E002125G 25 g
E0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0021100G Supplier TCI America Supplier No. E0021100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149-32-6
Molecular Formula C4H10O4
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004710
InChI Key UNXHWFMMPAWVPI-UHFFFAOYNA-N
Synonym erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
PubChem CID 222285
ChEBI CHEBI:17113
IUPAC Name butane-1,2,3,4-tetrol
SMILES OCC(O)C(O)CO

Specifications

Melting Point 120°C
Color White
Boiling Point 331°C
Quantity 100 g
Formula Weight 122.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material meso-Erythritol
EINECSNumber (2)-3651
RTECSNumber KF2000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.