Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelin Antibody (420411), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB313026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313026 Supplier Novus Biologicals Supplier No. FAB32652PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Mesothelin
Applications Flow Cytometry
Classification Monoclonal
Clone 420411
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Mesothelin, Glu296-Gly580, Accession # AAH09272
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.