Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesothelin Antibody (OTI8H7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N272651A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272651A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272651A647 Supplier Novus Biologicals Supplier No. NBP272651AF647

Mouse Monoclonal Antibody

Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Western Blot.

Specifications

Antigen Mesothelin
Applications Western Blot
Classification Monoclonal
Clone OTI8H7
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Gene Symbols MSLN
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 270-493 of human Mesothelin (NP_005814) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10232.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.