Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesp1 Antibody (2030B), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB295491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295491 Supplier Novus Biologicals Supplier No. FAB9219R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Mesp1 Monoclonal antibody specifically detects Mesp1 in Human samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen Mesp1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2030B
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1
Host Species Rabbit
Immunogen E. coli-dervived recombinant human MESP1, Met1-Gln85, Accession # Q9BRJ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 55897
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.