Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesp1 Antibody (939826), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB295499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295499 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295499 Supplier Novus Biologicals Supplier No. FAB92193MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mesp1 Monoclonal specifically detects Mesp1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Mesp1
Applications Western Blot
Classification Monoclonal
Clone 939826
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BHLHC5, bHLHc5MGC10676, Class C basic helix-loop-helix protein 5, mesoderm posterior 1 homolog (mouse), mesoderm posterior protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human MESP-1, Met1-Gln85, Accession # Q9BRJ9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 55897
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.