Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Mesulergine hydrochloride
GREENER_CHOICE

Catalog No. 164410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-441-0 10 mg
16-445-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-441-0 Supplier Tocris Bioscience™ Supplier No. 1644/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

5-HT2A and 5-HT2C antagonist; also dopamine receptor partial agonist

5-HT2A and 2C receptor antagonist (pA2 values are 9.1 for each receptor) and D2-like dopamine receptor partial agonist (Ki = 8 nM). Displays antiprolactin and antiparkinsonian effects in vivo.

Chemical Identifiers

CAS 72786-12-0
Molecular Formula C18H27ClN4O2S

Specifications

Quantity 10 mg
Formula Weight 398.95
Purity >99%
Molecular Weight (g/mol) 398.95
Chemical Name or Material Mesulergine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.