Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meta-Pneumovirus Antibody (HMPV123), Biotin, Novus Biologicals™
SDP

Catalog No. NB207257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB207257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB207257 Supplier Novus Biologicals Supplier No. NBP121631B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Meta-Pneumovirus Monoclonal antibody specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Meta-Pneumovirus
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone HMPV123
Conjugate Biotin
Formulation PBS
Gene Alias hMPV, metapneumovirus
Host Species Mouse
Immunogen Sub-type A hMPV virus isolate NCL03-4/145
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2799942
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.