Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meta-Pneumovirus Antibody (HMPV123), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP12631646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP12631646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP12631646 Supplier Novus Biologicals Supplier No. NBP121631JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Meta-Pneumovirus
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HMPV123
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias hMPV, metapneumovirus
Gene Symbols GLYG
Host Species Mouse
Immunogen Sub-type A hMPV virus isolate NCL03-4/145
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2799942
Test Specificity NBP1-21631 recognises the nucleoprotein of all sub-types (A1, A2, B1 and B2) of hMPV. NBP1-21631 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria.
Target Species Primate, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.