Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meta-Pneumovirus Antibody (HMPV57) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018466 0.025 mg
NB001897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018466 Supplier Novus Biologicals Supplier No. NBP1259410.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Meta-Pneumovirus
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HMPV57
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:80-1:160
Formulation PBS (pH 7.4)
Gene Alias hMPV, metapneumovirus
Gene Symbols GLYG
Host Species Mouse
Immunogen Sub-type A hMPV virus isolate NCL03-4/174
Quantity 0.025 mg
Gene ID (Entrez) 2799942
Test Specificity NBP1-25941 recognises human metapneumovirus (hMPV), a member of the Paramyxoviridae RNA virus family. Metapneumovirus was discovered in 2001 and is distributed worldwide. Infection in the upper respiratory tract is associated with mild illness, while lower respiratory tract infection can cause fever, cough, pneumonia, bronchiolitis and wheezing. Severe lower respiratory tract infection is most common in the very young, the very old and the immunosuppressed. Metapneumovirus is a common cause of respiratory infection.Phylogenetic analysis has shown two major genotypes, A and B, each containing two sub-divisions, suggesting that four distinct lineages circulate in the population with different lineages predominating in different years. NBP1-25941 recognises all sub-types (A1, A2, B1 and B2) of hMPV NBP1-25941 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria.The target antigen has not been determined for this antibody.
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.