Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meta-Pneumovirus Antibody (HMPV57) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $622.00
Specifications
Antigen | Meta-Pneumovirus |
---|---|
Clone | HMPV57 |
Concentration | 1 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1:80-1:160 |
Applications | Immunocytochemistry, Immunofluorescence |
Description
Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Meta-Pneumovirus | |
1 mg/mL | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
PBS (pH 7.4) | |
2799942 | |
Sub-type A hMPV virus isolate NCL03-4/174 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
HMPV57 | |
Immunocytochemistry/Immunofluorescence 1:80-1:160 | |
Monoclonal | |
Purified | |
Virus | |
hMPV, metapneumovirus | |
GLYG | |
IgG1 | |
NBP1-25941 recognises human metapneumovirus (hMPV), a member of the Paramyxoviridae RNA virus family. Metapneumovirus was discovered in 2001 and is distributed worldwide. Infection in the upper respiratory tract is associated with mild illness, while lower respiratory tract infection can cause fever, cough, pneumonia, bronchiolitis and wheezing. Severe lower respiratory tract infection is most common in the very young, the very old and the immunosuppressed. Metapneumovirus is a common cause of respiratory infection.Phylogenetic analysis has shown two major genotypes, A and B, each containing two sub-divisions, suggesting that four distinct lineages circulate in the population with different lineages predominating in different years. NBP1-25941 recognises all sub-types (A1, A2, B1 and B2) of hMPV NBP1-25941 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria.The target antigen has not been determined for this antibody. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title