Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meta-Pneumovirus Antibody (HMPV123) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
| Antigen | Meta-Pneumovirus |
|---|---|
| Clone | HMPV123 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:40-1:80, Immunoprecipitation |
| Applications | Immunocytochemistry, ELISA, Immunofluorescence, Flow Cytometry, Immunoprecipitation |
Description
Meta-Pneumovirus Monoclonal specifically detects Meta-Pneumovirus in Virus samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Meta-Pneumovirus | |
| 1.0 mg/mL | |
| Immunocytochemistry, ELISA, Immunofluorescence, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Virus, Primate | |
| 2799942 | |
| Sub-type A hMPV virus isolate NCL03-4/145 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| HMPV123 | |
| Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:40-1:80, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| hMPV, metapneumovirus | |
| GLYG | |
| IgG2a | |
| Protein G purified | |
| NBP1-21631 recognises the nucleoprotein of all sub-types (A1, A2, B1 and B2) of hMPV. NBP1-21631 does not cross-react with cell cultures infected with hRSV, influenzaviruses A and B, adenovirus, parainfluenza viruses 1, 2, 3 and 4b, mumps virus, measles virus, varicella-zoster virus, herpes simplex virus types 1 and 2, human cytomegalovirus, human herpesvirus type 6, ECHOvirus 19, Coxsachievirus B4, poliovirus types 1-3, HHV6 or uninfected HeLa, MA104, 3MK, LLC-MK, HEp2, MRC-5 and HSB-2 cells. It also does not react with sputum bacteria. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title