Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metaldehyde, 99%

Catalog No. ac271860050
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Catalog No. AC271860050

Supplier: Thermo Scientific Chemicals 271860050

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 108-62-3 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-62-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00071549
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
Synonym metaldehyde, metacetaldehyde, cekumeta, ariotox, halizan, metason, acetaldehyde, tetramer, slug-tox, corry's slug death, metaldehyd
PubChem CID 61021
ChEBI CHEBI:81931
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
SMILES CC1OC(OC(OC(O1)C)C)C

Specifications

Melting Point 246.0°C
Color White
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CHO)4
Quantity 5 g
Beilstein 01, 602
Merck Index 14, 5923
Solubility Information Solubility in water: 0.02 w/w % (20°C). Other solubilities: soluble in benzene,chloroform,sparingly soluble in alcohol,ether
Sublimation Point 0112.0°C
Formula Weight 176.21
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Metaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Flammable solid.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
We
EINECSNumber 203-600-2
RTECSNumber AB3042000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.