Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metallothionein-1A Antibody (UC1MT), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP1974930.05MG
Description
Metallothionein-1A Monoclonal specifically detects Metallothionein-1A in Human, Mouse, Rat, Equine, Fish, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Enzyme Immunoassay.Specifications
| metallothionein | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| Cross-linked rabbit liver Metallothionein I and II. | |
| 0.05 mg | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| UC1MT | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Enzyme Immunoassay | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human, Mouse, Rat, Equine, Fish, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction