Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metallothionein-1A Antibody (UC1MT), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$246.61 - $776.74
Specifications
| Antigen | metallothionein |
|---|---|
| Clone | UC1MT |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Enzyme Immunoassay |
| Applications | Immunohistochemistry (Paraffin), Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018787
![]() |
Novus Biologicals
NBP1974930.025MG |
0.025 mg |
Each for $246.61
|
|
|||||
NBP19749305
![]() |
Novus Biologicals
NBP1974930.05MG |
0.05 mg |
Each for $443.24
|
|
|||||
NBP19749302
![]() |
Novus Biologicals
NBP1974930.2MG |
0.2 mg |
Each for $776.74
|
|
|||||
Description
Metallothionein-1A Monoclonal specifically detects Metallothionein-1A in Human, Mouse, Rat, Equine, Fish, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Enzyme Immunoassay.Specifications
| metallothionein | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Enzyme Immunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| IgG1 | |
| Protein G purified |
| UC1MT | |
| Immunohistochemistry (Paraffin), Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Equine, Rat, Rabbit, Fish, Mouse, Human | |
| Cross-linked rabbit liver Metallothionein I and II. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title