Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

metallothionein Antibody (2B5), Novus Biologicals™
SDP

Catalog No. NB171886 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171886 Supplier Novus Biologicals Supplier No. NBP318262
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

metallothionein Monoclonal antibody specifically detects metallothionein in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen metallothionein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B5
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:800
Formulation PBS pH 7.4, 50% glycerol
Host Species Mouse
Immunogen Recombinant full length Pseudomonas aeruginosa Metallothionein protein
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.