Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MT1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$188.00 - $536.00
Specifications
| Antigen | MT1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5121779
|
Invitrogen™
PA5121779 |
20 μL |
Each for $188.00
|
|
|||||
PIPA5121778
![]() |
Invitrogen™
PA5121778 |
100 μL |
Each for $536.00
|
|
|||||
Description
This gene is a member of the metallothionein family of genes. Proteins encoded by this gene family are low in molecular weight, are cysteine-rich, lack aromatic residues, and bind divalent heavy metal ions. The conserved cysteine residues co-ordinate metal ions using mercaptide linkages. These proteins act as anti-oxidants, protect against hydroxyl free radicals, are important in homeostatic control of metal in the cell, and play a role in detoxification of heavy metals. Disruption of two metallothionein genes in mouse resulted in defects in protection against heavy metals, oxidative stress, immune reactions, carcinogens, and displayed obesity. MT1A (Metallothionein 1A) is a Protein Coding gene. Diseases associated with MT1A include Menkes Disease and Deficiency Anemia. Among its related pathways are Metal ion SLC transporters and Cellular responses to stimuli. An important paralog of this gene is MT2A.Specifications
| MT1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P04731 | |
| 4489 | |
| Met1-Ala61 (Accession P04731), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MT1A |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Metallothionein; Metallothionein-1 | |
| MT1A | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title