Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metapneumovirus Nucleoprotein Antibody (hMPV123) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB413834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413834 0.025 mL
NBP253104 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413834 Supplier Novus Biologicals Supplier No. NBP2531040.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Metapneumovirus Nucleoprotein Monoclonal specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Metapneumovirus Nucleoprotein
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone hMPV123
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Host Species Mouse
Immunogen Human MPV strain NCL-145
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Identification and location of human metapneumovirus (hMPV) of both types A and B.
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.