Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metapneumovirus Nucleoprotein Antibody (hMPV123) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $501.50
Specifications
Antigen | Metapneumovirus Nucleoprotein |
---|---|
Clone | hMPV123 |
Concentration | 1.0 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Metapneumovirus Nucleoprotein Monoclonal specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
Metapneumovirus Nucleoprotein | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
Human MPV strain NCL-145 | |
Primary | |
Identification and location of human metapneumovirus (hMPV) of both types A and B. |
hMPV123 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
IgG2a | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title