Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metapneumovirus Phosphoprotein Antibody (hMPV33), HRP, Novus Biologicals™
SDP

Catalog No. NBP250506H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250506H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250506H Supplier Novus Biologicals Supplier No. NBP250506H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Metapneumovirus Phosphoprotein Monoclonal specifically detects Metapneumovirus Phosphoprotein in Virus samples. It is validated for ELISA.

Specifications

Antigen Metapneumovirus Phosphoprotein
Applications ELISA
Classification Monoclonal
Clone hMPV33
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Host Species Mouse
Immunogen Human hMPV strain NCL-174
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Identifies Metapneumovirus nucleoprotein of both types A and B.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.