Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanesulfonic acid, 98%

Catalog No. p-4259039
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass Bottle
Plastic bottle

CAS: 75-75-2 | CH4O3S | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Melting Point 19.0°C
Color Yellow to Brown
Density 1.4810g/mL
Boiling Point 167.0°C (10.0 mmHg)
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4252 to 1.4315
Linear Formula CH3SO3H
Quantity 1 L
Merck Index 15,6026
Specific Gravity 1.481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C)
Viscosity 11.6 mPa.s (25°C)
Formula Weight 96.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
D
EINECSNumber 200-898-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.