Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanesulfonic Acid 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 122°C |
| Quantity | 100 g |
| UN Number | 2586 |
| Formula Weight | 96.10 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Methanesulfonic Acid [for HPLC] |
Safety and Handling
| EINECSNumber | (2)-1582 |
| RTECSNumber | PB1140000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction