Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
SDP

Catalog No. r482900025c Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
25 mL
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Hard Pack
Metal Can
Natural Poly Bottle

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Boiling Point 64.5°C
Quantity 25 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0003 meq/g
Water max. 0.1%
Synonym methyl alcohol
Percent Purity min. 99.8%
Grade ACS
Chemical Name or Material Methanol, Absolute
Show More Show Less
ShelfLife 36 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.