Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Anhydrous, DriSolv™, Septum-Sealed, MilliporeSigma™

Catalog No. p-3797322 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 mL
1 L

Packed under Nitrogen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 100 mL
Physical Form Liquid Free From Particulates
Residue after Evaporation 1ppm max.
Assay Percent Range 99.8% min.
Linear Formula CH3OH
Water 50ppm max.
Formula Weight 32.04
Chemical Name or Material Methanol, Septum-sealed
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.