Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Pressure Bottom Poly Bottle
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Quantity | 4 L |
| Flash Point | 12.00°C (53.6°F) |
| Packaging | Pressure Bottom Poly Bottle |
| Specific Gravity | 1L = 0.80kg |
| Formula Weight | 32.04 |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction