Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Catalog No. p-7073736 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2 L
2.5 L
4 x 2 L
4 x 2.5 L
4 x 4 L
6 x 1 L

Catalog No. 60-009-36

Supplier: Honeywell Chemicals 348606X1L

Only null left
Add to Cart
Add to Cart

Product filtered through a 0.2μm filter

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Density 0.791g/mL at 25°C
Boiling Point 64.7°C
Quantity 1 L
Refractive Index n20/D 1.329
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C)
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV
Chemical Name or Material Methanol, for HPLC
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
Precautionary Statement H225-H301 + H311 + H331-H370
EINECSNumber 200-659-6
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.