Learn More
Methanol (HPLC), Fisher Chemical PROMO
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | -98°C | 
| Absorbance | 0.025 AU Max at 254 nm, 0.15 AU max at 230 nm, 0.30 AU max at 220 nm, 1.00 AU max at 205 nm | 
| Color | Colorless | 
| Boiling Point | 64.7°C | 
| Quantity | 4 L | 
| Identification | Pass Test | 
| Packaging | Glass Bottle | 
| Physical Form | Liquid | 
| Assay Percent Range | Assay: 99.9% min by GC | 
| Residue after Evaporation | 3 ppm Max | 
| Show More | 
Safety and Handling
                        
                        
                            
                        
                        | Hazard Statement | DANGER! | 
| Precautionary Statement | Emergency Overview: Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and eyes. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. NFPA Health:1 Flammability:3 Instability:0  | 
                                
| DOTInformation | DOT Class 3, : Flammable Liquid | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.