Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, HPLC Grade, MilliporeSigma™

Catalog No. mmx0475p6 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle (Pre-packed)

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.792 g/cc at 20°C
Boiling Point 64.5°C (1013 hPa)
Quantity 1 L
Flash Point 10°C
Ignition Point 455°C (55°F)
Odor Characteristic
Packaging Glass Bottle (Pre-packed)
Physical Form Clear liquid free from particulates
Residue after Evaporation 2ppm
Titratable Acid 0.0003meq/g
Vapor Pressure 128 hPa (20°C)
Viscosity 0.597 mPa.s at 20°C
Water 0.05%
Formula Weight 32.04
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Methanol
Show More Show Less
Hazard Category Danger
Hazard Statement H225 Highly flammable liquid and vapor.
H301 + H311 + H331 Toxic if swallowed, in contact with skin or if inhaled.H370 Causes damage to organs (Eyes)
Precautionary Statement P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P233 Keep container tightly closed. P280 Wear protective gloves/ protective clothing. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P304 + P340 IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. P308 + P310 IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician. P403 + P235 Store in a well-ventilated place. Keep cool.
EINECSNumber 200-659-6
RTECSNumber PC1400000
Recommended Storage Store at room temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.