Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, LC-MS CHROMASOLV™, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities

Specifications
Specifications
Density | 0.791g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.36 |
Flash Point | 9°C (48.2°F) |
Ignition Point | 725°F |
Linear Formula | CH3OH |
Vapor Pressure | 410 mmHg (50°C), 97.68 mmHg (20°C) |
UN Number | UN1230 |
Beilstein | 1098229 |
Formula Weight | 32.04g/mol |
Show More |
Safety and Handling
missing translation for 'healthHazard2' | P210-P260-P280-P301 + P310-P311 |
RTECSNumber | PC1400000 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction